Learn More
Medchemexpress LLC D-Xylulose-2-13C | 131771-47-6 | 99.5% | 151.12 | 50 MG
SDP
Catalog No. 5000454030
Encompass_Preferred
D-Xylulose-2-13C is the 13C labeled D-Xylulose. D-xylulose is a precursor of the pentiol D-arabi. This product is for research use only and not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54030 Supplier Medchemexpress LLC Supplier No. HYW010256S150MG
D-Xylulose-2-13C is the 13C labeled D-Xylulose. D-xylulose is a precursor of the pentiol D-arabi. This product is for research use only and not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.