Learn More
TARGETMOL CHEMICALS INC D-Xylulose 25MG
SDP
Catalog No. 502880272
Encompass_Preferred
Also available in 1 mg, 50 mg and bulk. Please contact Fisher for quotes. D-Xylulose is converted from xylitol by the enzyme NAD+-linked xylitol dehydrogenase in the glucuronate pathway, the most important xylitol-handling metabolic pathway in mammals.
Catalog No. 50-288-0272 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ019025MG
Also available in 1 mg, 50 mg and bulk. Please contact Fisher for quotes. D-Xylulose is converted from xylitol by the enzyme NAD+-linked xylitol dehydrogenase in the glucuronate pathway, the most important xylitol-handling metabolic pathway in mammals.