Learn More
Medchemexpress LLC D-xylulose 5-phosphate sodium | 105931-44-0 | MFCD00070073 | 90.0% | 252.09 g/mol | C5H10NaO8P | 1mg
SDP
Supplier:  Medchemexpress LLC HYW7123271MG
Encompass_Preferred
D-Xylulose 5-phosphate (sodium) is a pentose phosphate ester and an essential intermediate metabolite in the pentose phosphate pathway (PPP) and the non-oxidative phase of the pentose phosphate pathway D-xylulose-5-phosphate can be efficiently synthesized through the phosphorylation of D-xylulose catalyzed by D-xylulokinase (XKS1 from Saccharomyces cerevisiae) with ATP regeneration facilitated by the phosphoenolpyruvate (PEP)/pyruvate kinase (PK) system[1]
Catalog No. 50-002-70154