Learn More
Montana Molecular Green Ratiometric Cilia-Targeted cADDis cAMP Sensor Kit D0211G, 10mL, CMV
SDP
Catalog No. 501631913
Encompass_Preferred
Measure real-time cAMP signaling kinetics in the primary cilia with this live cell fluorescent assay. Add cADDis to your cells and measure robust signals the next day on a fluorescence imaging system. Decreasing green fluorescence indicates ciliary cAMP increase. Constitutively expressed cytosolic red fluorescence can be used for ratiometric measurements. This assay is genetically encoded and packaged in BacMam for efficient delivery to cilia in most cell types, including many primary and iPSC-derived cells.
Catalog No. 50-163-1913 Supplier Montana Molecular Supplier No. D0211G010
Measure real-time cAMP signaling kinetics in the primary cilia with this live cell fluorescent assay. Add cADDis to your cells and measure robust signals the next day on a fluorescence imaging system. Decreasing green fluorescence indicates ciliary cAMP increase. Constitutively expressed cytosolic red fluorescence can be used for ratiometric measurements. This assay is genetically encoded and packaged in BacMam for efficient delivery to cilia in most cell types, including many primary and iPSC-derived cells.