Learn More
Montana Molecular Green Downward cADDis cAMP sensor Big Sky BacMam 5mL
SDP
Catalog No. 502815247
Encompass_Preferred
Detect cAMP signaling kinetics in living cells with the Green Downward cADDis fluorescent cAMP assay. Decrease in fluorescence indicates increase in cAMP. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This is an update to product D0200G, with improved expression efficiency from Big Sky BacMam.
Catalog No. 50-281-5247 Supplier Montana Molecular Supplier No. D0250G005
Detect cAMP signaling kinetics in living cells with the Green Downward cADDis fluorescent cAMP assay. Decrease in fluorescence indicates increase in cAMP. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This is an update to product D0200G, with improved expression efficiency from Big Sky BacMam.