Learn More
Montana Molecular Green Downward DAG sensor Big Sky BacMam 5mL
SDP
Catalog No. 502815248
Encompass_Preferred
Detect DAG or Gq signaling kinetics in living cells with the Green Downward fluorescent DAG assay. Decrease in fluorescence indicates increase in DAG. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This is an update to product D0300G, with improved expression efficiency from Big Sky BacMam.
Catalog No. 50-281-5248 Supplier Montana Molecular Supplier No. D0350G005
Detect DAG or Gq signaling kinetics in living cells with the Green Downward fluorescent DAG assay. Decrease in fluorescence indicates increase in DAG. Packaged in Big Sky BacMam, for enhanced expression in HEKs, CHOs, and many other mammalian cell types and iPSCs. This is an update to product D0300G, with improved expression efficiency from Big Sky BacMam.