Learn More
Montana Molecular Green Downward GENIe cGMP Sensor Kit D0800G, 30mL, CMV
SDP
Catalog No. 501631936
Encompass_Preferred
Measure cyclic GMP (cGMP) signaling dynamics and ascertain the specificity of PDE inhibitors or SNP activators with this live cell fluorescent assay. Add GENIe to your cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. Decreasing green fluorescence indicates cGMP increase. The assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. Kit includes soluble guanylate cyclase..
Catalog No. 50-163-1936 Supplier Montana Molecular Supplier No. D0800G030
Measure cyclic GMP (cGMP) signaling dynamics and ascertain the specificity of PDE inhibitors or SNP activators with this live cell fluorescent assay. Add GENIe to your cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. Decreasing green fluorescence indicates cGMP increase. The assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. Kit includes soluble guanylate cyclase..