Learn More
Alkali Scientific D1000 Handheld Homogenizer
SDP
Catalog No. 503029526
Encompass_Preferred
Alkalis Hand-held Homogenizer with 5mm and 7mm Generator Heads the D1000 is designed to rapidly homogenize emulsify suspend and disrupt biological samples Ideal for use with microtubes the homogenizer works well with samples from 0 1ml to 250ml (depending upon the generator probe used) The D1000 is a rotor stator homogenizer and works by drawing the sample in liquid into the generator probe with the variable speed rotor and then forcing it out through the slots in the stator This achieves mechanical shearing of the sample As the sample is repeatedly drawn up and forced through the stator additional shearing takes place Most samples are completely homogenized in 30 seconds or less A powerful 130W motor drives the rotor at speeds from 8500 to 30000 rpm Speed is adjustable in sixsteps to meet sample processing needs A separate on/off switch allows the homogenizer to be left at the appropriate setting Generator probes are available in four sizes The 5mm and 7mm diameter probes h
Catalog No. 50-302-9526 Supplier Alkali Scientific Supplier No. RS7410
Alkalis Hand-held Homogenizer with 5mm and 7mm Generator Heads the D1000 is designed to rapidly homogenize emulsify suspend and disrupt biological samples Ideal for use with microtubes the homogenizer works well with samples from 0 1ml to 250ml (depending upon the generator probe used) The D1000 is a rotor stator homogenizer and works by drawing the sample in liquid into the generator probe with the variable speed rotor and then forcing it out through the slots in the stator This achieves mechanical shearing of the sample As the sample is repeatedly drawn up and forced through the stator additional shearing takes place Most samples are completely homogenized in 30 seconds or less A powerful 130W motor drives the rotor at speeds from 8500 to 30000 rpm Speed is adjustable in sixsteps to meet sample processing needs A separate on/off switch allows the homogenizer to be left at the appropriate setting Generator probes are available in four sizes The 5mm and 7mm diameter probes h