Learn More
Agilent Technologies D1000 HIGH SENS DNA SCREENTAPE
Catalog No. NC2192415
Encompass
High Sensitivity D1000 Ladder. For the high sensitivity analysis of DNA from 35 to 1000 bp. Ladder only.
This item is not returnable. View return policy
Catalog No. NC2192415 Supplier Agilent Technologies Supplier No. 50675587
High Sensitivity D1000 Ladder. For the high sensitivity analysis of DNA from 35 to 1000 bp. Ladder only.