Learn More
Agilent Technologies D1000 Ladder
Catalog No. NC1844198
Encompass
For the analysis of DNA from 35 to 1000 bp. Ladder only.
Catalog No. NC1844198 Supplier Agilent Technologies Supplier No. 50675586
For the analysis of DNA from 35 to 1000 bp. Ladder only.