Learn More
Agilent Technologies DNA ScreenTape Analysis, 1 each
Catalog No. NC1883382
Encompass
D1000 Reagents. For the analysis of DNA from 35 to 1000 bp. Includes ladder and sample buffer sufficient for 112 samples.
Catalog No. NC1883382 Supplier Agilent Technologies Supplier No. 50675583
D1000 Reagents. For the analysis of DNA from 35 to 1000 bp. Includes ladder and sample buffer sufficient for 112 samples.