Learn More
Agilent Technologies D1000 Sample Buffer
Catalog No. NC1844201
Encompass
For the analysis of DNA from 35 to 1000 bp. Sample buffer only.
Catalog No. NC1844201 Supplier Agilent Technologies Supplier No. 50675602
For the analysis of DNA from 35 to 1000 bp. Sample buffer only.