Learn More
Thomas Scientific SCILOGEX D1008 Mini Centrifuge, Green Lid, c/w 8 x 2ml rotor & 2 x PCR8 rotor, 100-240V, 50/60Hz
SDP
Catalog No. NC1343938
Encompass_Preferred
Ideal for quick spin downs of microtubes and PCR tubes 8-Place 1.5/2.0ml Microtube rotor 7000rpm / 2680xg Features Four color options, green, yellow, blue and pink Maximum speed 7000rpm/2680xg Supplied with an 8 place 1.5/2.0ml microtube rotor and PCR rotor Compact design Whisper quiet operation Ideal for quick spin downs of microtubes and PCR tubes Simply close the lid for acceleration and close the lid for deceleration, or use the on/off switch Universal voltage 2 year manufacturer warranty
Catalog No. NC1343938 Supplier Thomas Scientific Supplier No. 1205R42
Ideal for quick spin downs of microtubes and PCR tubes 8-Place 1.5/2.0ml Microtube rotor 7000rpm / 2680xg Features Four color options, green, yellow, blue and pink Maximum speed 7000rpm/2680xg Supplied with an 8 place 1.5/2.0ml microtube rotor and PCR rotor Compact design Whisper quiet operation Ideal for quick spin downs of microtubes and PCR tubes Simply close the lid for acceleration and close the lid for deceleration, or use the on/off switch Universal voltage 2 year manufacturer warranty