Learn More
Alkali Scientific Alkali Scientific™ D1032-01 Prefilled 2.0ml tubes, Zirconium Beads, 0.1mm Triple-Pure - High Impact, 50/PK
SDP
Catalog No. 503027635
Encompass_Preferred
These Alkali Scientific™ D1032-01 Prefilled 2.0ml tubes are prefilled with an appropriate amount of beads for bead homogenization. Tubes are ready to use – beads are acid washed and heat treated to remove contaminants and render them nuclease and protease free. Suitable for homogenization of bacteria, fungi, spores, molds and small yeasts. Compatible with BeadBug™ Microtube homogenizers and all common bead homogenizers. The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease. The stainless steel beads are acid-washed as well. All tubes are non-skirted with screw-cap.
Catalog No. 50-302-7635 Supplier Alkali Scientific Supplier No. RS70125
These Alkali Scientific™ D1032-01 Prefilled 2.0ml tubes are prefilled with an appropriate amount of beads for bead homogenization. Tubes are ready to use – beads are acid washed and heat treated to remove contaminants and render them nuclease and protease free. Suitable for homogenization of bacteria, fungi, spores, molds and small yeasts. Compatible with BeadBug™ Microtube homogenizers and all common bead homogenizers. The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease. The stainless steel beads are acid-washed as well. All tubes are non-skirted with screw-cap.