Learn More
Alkali Scientific Alkali Scientific™ (D1033-28) Prefilled 2.0ml tubes, Stainless Steel, 2.8mm Acid Washed, 50/PK
SDP
Catalog No. 503028921
Encompass_Preferred
These Prefilled 2.0ml tubes, Stainless Steel, 2.8mm Acid Washed, and come with standard acid-washed glass beads, triple-pure zirconium beads, or stainless steel beads. The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease. The stainless steel beads are acid-washed as well. All tubes are non-skirted with screw-cap. Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.
Catalog No. 50-302-8921 Supplier Alkali Scientific Supplier No. RS70130
These Prefilled 2.0ml tubes, Stainless Steel, 2.8mm Acid Washed, and come with standard acid-washed glass beads, triple-pure zirconium beads, or stainless steel beads. The triple-pure beads are molecular biology grade, and are acid washed, heat treated and tested free of nuclease. The stainless steel beads are acid-washed as well. All tubes are non-skirted with screw-cap. Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.