Learn More
Alkali Scientific Alkali Scientific™ (D1033-30G) Prefilled 2.0ml tubes, 0.1mm Silica, 1.4 mm Zirconium & 4mm Silica Beads, 50pk
SDP
Catalog No. 503029345
Encompass_Preferred
Prefilled 2.0ml tubes contain a mix of 0.1mm Silica, 1.4 mm diameter Zirconium & 4mm diameter Silica Beads. Useful for homogenization of soil, sludge, feces and environmental samples. Beads are acid washed and heat treated to remove contaminants and render them nuclease and protease free. Supplied in packs of 50 tubes. Tubes are suitable for use in most bead homogenizers including the BeadBug 3, BeadBug 6 and BeadBlaster 24.
Catalog No. 50-302-9345 Supplier Alkali Scientific Supplier No. RS70129MX
Prefilled 2.0ml tubes contain a mix of 0.1mm Silica, 1.4 mm diameter Zirconium & 4mm diameter Silica Beads. Useful for homogenization of soil, sludge, feces and environmental samples. Beads are acid washed and heat treated to remove contaminants and render them nuclease and protease free. Supplied in packs of 50 tubes. Tubes are suitable for use in most bead homogenizers including the BeadBug 3, BeadBug 6 and BeadBlaster 24.