Learn More
Grainger NORTON DOOR CONTROLS Door Closer, Parallel Mount, Grade 1, 13 5/8 in., 1 ea.
Catalog No. 502792849
Encompass_Preferred
This heavy-duty, Grade 1 institutional door closer is designed for parallel arm mounting, offering a secure and durable solution for both interior and exterior doors. Featuring a non-handed, hydraulic operation, this unit is engineered to support doors up to 250 lb. and provides reliable, adjustable performance in high-traffic applications.
  • Features a durable cast aluminum housing with a dark bronze painted finish.
  • Includes staked valves for reliable performance.
  • Tri-packed design allows for versatile installation.
  • ANSI/BHMA A156.4 and A117.1 compliant, and UL/cUL listed for fire safety.
  • Suitable for fire-rated openings up to 3 hr.
  • Designed with a 110° maximum door opening angle.
Catalog No. 50-279-2849 Supplier Grainger Supplier No. 1ABJ7
This heavy-duty, Grade 1 institutional door closer is designed for parallel arm mounting, offering a secure and durable solution for both interior and exterior doors. Featuring a non-handed, hydraulic operation, this unit is engineered to support doors up to 250 lb. and provides reliable, adjustable performance in high-traffic applications.
  • Features a durable cast aluminum housing with a dark bronze painted finish.
  • Includes staked valves for reliable performance.
  • Tri-packed design allows for versatile installation.
  • ANSI/BHMA A156.4 and A117.1 compliant, and UL/cUL listed for fire safety.
  • Suitable for fire-rated openings up to 3 hr.
  • Designed with a 110° maximum door opening angle.