Learn More
Grainger Norton 7500 Series Door Closer, Grade 1, Heavy Duty, 1 ea
Catalog No. 502793045
Encompass_Preferred
The 7500 Series heavy-duty door closer provides reliable, versatile operation for a wide range of commercial, public, and private applications. Designed for durability in high-traffic environments, this non-handed unit features a cast aluminum body and precision rack-and-pinion mechanism to ensure smooth, consistent closing. It is certified to ANSI/BHMA A156.4 Grade 1 standards and is suitable for use on fire and smoke protection doors.
  • Heavy-duty cast aluminum body for long-lasting durability
  • Universal tri-style mounting supports regular, top jamb, or parallel arm installations
  • Non-handed design for flexible left or right-handed door application
  • Adjustable spring sizes 1 to 6 allow for customizable closing force
  • Variable closing and latching speeds provide controlled operation
  • Tested for use in fire and smoke protection doors according to UL10C
  • Adjustable back check and dela
Catalog No. 50-279-3045 Supplier Grainger Supplier No. 1ABJ8
The 7500 Series heavy-duty door closer provides reliable, versatile operation for a wide range of commercial, public, and private applications. Designed for durability in high-traffic environments, this non-handed unit features a cast aluminum body and precision rack-and-pinion mechanism to ensure smooth, consistent closing. It is certified to ANSI/BHMA A156.4 Grade 1 standards and is suitable for use on fire and smoke protection doors.
  • Heavy-duty cast aluminum body for long-lasting durability
  • Universal tri-style mounting supports regular, top jamb, or parallel arm installations
  • Non-handed design for flexible left or right-handed door application
  • Adjustable spring sizes 1 to 6 allow for customizable closing force
  • Variable closing and latching speeds provide controlled operation
  • Tested for use in fire and smoke protection doors according to UL10C
  • Adjustable back check and dela