Learn More
ATCC D10.G4.1; T Lymphocyte; Mouse (Mus musculus)
Catalog No. 502381774
Encompass
D10.G4.1 is a helper t lymphocyte from a lymph node of a male mouse. These cells can also be used as a reliable and sensitive assay for IL-1 (interleukin-1) response, since D10 cells will proliferate only minimally to con A in the absence of IL-1 or feeder cells.
Catalog No. 50-238-1774 Supplier ATCC Supplier No. TIB224
D10.G4.1 is a helper t lymphocyte from a lymph node of a male mouse. These cells can also be used as a reliable and sensitive assay for IL-1 (interleukin-1) response, since D10 cells will proliferate only minimally to con A in the absence of IL-1 or feeder cells.