Learn More
Grainger Norton Door Closer, Top Frame, Grade 1, Heavy Duty, 1 ea
Catalog No. 502793157
Encompass_Preferred
This heavy-duty, surface-mounted door closer is engineered for architectural openings in moderate- to high-traffic environments. Featuring a versatile, non-handed design, it is compatible with interior and exterior doors. The unit utilizes a rack and pinion mechanism with a multi-sized adjustable spring power (sizes 1 through 6) to provide precise, consistent control throughout the full opening and closing cycle. It is constructed from a specialized aluminum alloy for durability and is certified to meet Grade 1 performance standards.
  • Provides independent regulation of sweep and latch speeds via two non-critical, hex-key adjusted valves
  • Includes adjustable backcheck intensity to cushion the door during opening
  • Designed for 3 hr fire-rated doors and complies with ANSI/BHMA A156.4 requirements
  • Tri-Style® packaging supports regular, top jamb, and parallel arm mounting configurations
  • Features
Catalog No. 50-279-3157 Supplier Grainger Supplier No. 1ABK1
This heavy-duty, surface-mounted door closer is engineered for architectural openings in moderate- to high-traffic environments. Featuring a versatile, non-handed design, it is compatible with interior and exterior doors. The unit utilizes a rack and pinion mechanism with a multi-sized adjustable spring power (sizes 1 through 6) to provide precise, consistent control throughout the full opening and closing cycle. It is constructed from a specialized aluminum alloy for durability and is certified to meet Grade 1 performance standards.
  • Provides independent regulation of sweep and latch speeds via two non-critical, hex-key adjusted valves
  • Includes adjustable backcheck intensity to cushion the door during opening
  • Designed for 3 hr fire-rated doors and complies with ANSI/BHMA A156.4 requirements
  • Tri-Style® packaging supports regular, top jamb, and parallel arm mounting configurations
  • Features