Learn More
Grainger Norton Door Controls Door Closer, Top Frame, Grade 1, 1 ea
Catalog No. 502792543
Encompass_Preferred
This heavy-duty door closer is designed for top frame mounting on the push side of the door, offering a reliable solution for areas with limited door-mounted space. Engineered for high-traffic environments, it features adjustable spring tension and a non-handed design for versatile installation.
  • Performance rated as grade 1 heavy duty
  • Supports a maximum door opening angle of 110°
  • Features a 3 hr fire rating
  • ADA compliant and non-handed for universal application
  • Designed for exterior use
  • Equipped with hydraulic energy source for consistent operation
Catalog No. 50-279-2543 Supplier Grainger Supplier No. 1ABK2
This heavy-duty door closer is designed for top frame mounting on the push side of the door, offering a reliable solution for areas with limited door-mounted space. Engineered for high-traffic environments, it features adjustable spring tension and a non-handed design for versatile installation.
  • Performance rated as grade 1 heavy duty
  • Supports a maximum door opening angle of 110°
  • Features a 3 hr fire rating
  • ADA compliant and non-handed for universal application
  • Designed for exterior use
  • Equipped with hydraulic energy source for consistent operation