Learn More
Grainger Norton Door Controls Door Closer, Heavy Duty, Hydraulic, Regular Arm, 1 ea
Catalog No. 502792900
Encompass_Preferred
This heavy-duty, manual, hydraulic door closer is designed for both interior and exterior applications. It features a robust cast aluminum housing and a regular arm configuration for reliable performance in institutional settings.
  • Performance grade 1 construction
  • Adjustable spring size 2-4
  • Compatible with interior doors up to 48 in. wide and exterior doors up to 36 in. wide
  • Supports a maximum door weight of 250 lb.
  • Non-hold open design with 180° maximum door opening
  • UL and cUL listed with a 3 hr. fire rating per UL10C
  • Dark bronze painted finish
  • Left-handed orientation
Catalog No. 50-279-2900 Supplier Grainger Supplier No. 1ABF6
This heavy-duty, manual, hydraulic door closer is designed for both interior and exterior applications. It features a robust cast aluminum housing and a regular arm configuration for reliable performance in institutional settings.
  • Performance grade 1 construction
  • Adjustable spring size 2-4
  • Compatible with interior doors up to 48 in. wide and exterior doors up to 36 in. wide
  • Supports a maximum door weight of 250 lb.
  • Non-hold open design with 180° maximum door opening
  • UL and cUL listed with a 3 hr. fire rating per UL10C
  • Dark bronze painted finish
  • Left-handed orientation