Learn More
Grainger Norton Traditional Surface Closer, Aluminum Alloy, Regular Arm, 1 ea
Catalog No. 502792872
Encompass_Preferred
The traditional surface door closer offers a robust, heavy-duty solution for various door environments. Engineered for versatility, it features an adjustable clock coil spring that accommodates a wide range of door sizes and power requirements. The unit provides adjustable sweep, latch, and backcheck cushioning, allowing for precise control over door closing speed and operation based on site conditions. It is constructed from a durable, cast aluminum alloy and designed for long-term, high-frequency use in demanding applications.
  • Performance grade 1 for heavy-duty applications
  • Adjustable spring power and closing speeds
  • Rack-and-pinion operation for smooth transmission of power
  • Compatible with both interior and exterior doors
  • Three-hour fire rating for safety compliance
  • UL and cUL listed
Catalog No. 50-279-2872 Supplier Grainger Supplier No. 1ABN2
The traditional surface door closer offers a robust, heavy-duty solution for various door environments. Engineered for versatility, it features an adjustable clock coil spring that accommodates a wide range of door sizes and power requirements. The unit provides adjustable sweep, latch, and backcheck cushioning, allowing for precise control over door closing speed and operation based on site conditions. It is constructed from a durable, cast aluminum alloy and designed for long-term, high-frequency use in demanding applications.
  • Performance grade 1 for heavy-duty applications
  • Adjustable spring power and closing speeds
  • Rack-and-pinion operation for smooth transmission of power
  • Compatible with both interior and exterior doors
  • Three-hour fire rating for safety compliance
  • UL and cUL listed