Learn More
Grainger Norton Pot-Belly Door Closer, Cast Aluminum, Regular Arm, Right-Handed, 1 ea
Catalog No. 502792679
Encompass_Preferred
This heavy-duty hydraulic door closer is designed for right-handed manual operation in both interior and exterior institutional settings. Constructed from cast aluminum with a dark bronze painted finish, this Grade 1 performance device ensures reliable control for doors weighing up to 250 lb. The unit features an adjustable spring size ranging from 5 to 6 and is capable of a 180° opening angle.
  • Performance grade 1 construction for institutional use
  • Adjustable spring size 5 to 6
  • Non hold open operation
  • 3 hr. UL10C fire rating
  • Compatible with interior doors up to 48 in. and exterior doors up to 44 in. wide
  • Regular arm mounting style
Catalog No. 50-279-2679 Supplier Grainger Supplier No. 1ABN9
This heavy-duty hydraulic door closer is designed for right-handed manual operation in both interior and exterior institutional settings. Constructed from cast aluminum with a dark bronze painted finish, this Grade 1 performance device ensures reliable control for doors weighing up to 250 lb. The unit features an adjustable spring size ranging from 5 to 6 and is capable of a 180° opening angle.
  • Performance grade 1 construction for institutional use
  • Adjustable spring size 5 to 6
  • Non hold open operation
  • 3 hr. UL10C fire rating
  • Compatible with interior doors up to 48 in. and exterior doors up to 44 in. wide
  • Regular arm mounting style