Learn More
Medchemexpress LLC D13-9001 | 957471-96-4 | 99.9% | 693.78 g·mol⁻¹ | C31H39N11O6S | 10MG
SDP
Catalog No. 5000239301
Encompass_Preferred
D13-9001 is a small-molecule bacterial efflux pump inhibitor that selectively targets RND-family transporters AcrB and MexB. It binds AcrB and MexB with reported KD values of 1.15 μM and 3.57 μM, respectively, and demonstrates antibiotic activity in vitro. The compound is supplied for research use in multiple pack sizes, including a 10 mg option.
  • Inhibits AcrB and MexB efflux pump subunits.
  • Measured KD values of 1.15 μM (AcrB) and 3.57 μM (MexB).
  • Molecular weight 693.78 g·mol⁻¹ and formula C31H39N11O6S.
  • High purity: 99.85% (typical).
  • Suitable for in vitro antibiotic activity and efflux pump studies.
  • Available in small research pack sizes, including 10 mg.
Catalog No. 50-002-39301 Supplier Medchemexpress LLC Supplier No. HY12481910MG
D13-9001 is a small-molecule bacterial efflux pump inhibitor that selectively targets RND-family transporters AcrB and MexB. It binds AcrB and MexB with reported KD values of 1.15 μM and 3.57 μM, respectively, and demonstrates antibiotic activity in vitro. The compound is supplied for research use in multiple pack sizes, including a 10 mg option.
  • Inhibits AcrB and MexB efflux pump subunits.
  • Measured KD values of 1.15 μM (AcrB) and 3.57 μM (MexB).
  • Molecular weight 693.78 g·mol⁻¹ and formula C31H39N11O6S.
  • High purity: 99.85% (typical).
  • Suitable for in vitro antibiotic activity and efflux pump studies.
  • Available in small research pack sizes, including 10 mg.