Learn More
Medchemexpress LLC D13-9001 | 957471-96-4 | 99.9% | 693.78 g/mol | C31H39N11O6S | 50MG
SDP
Catalog No. 5000237107
Encompass_Preferred
D13-9001 is a small-molecule efflux pump inhibitor used as a research tool to probe AcrB- and MexB-mediated antibiotic efflux. It is intended for laboratory research use only and is supplied as a solid with high reported purity and documented physicochemical properties.
Catalog No. 50-002-37107 Supplier Medchemexpress LLC Supplier No. HY12481950MG
Add to Cart
Add to Cart
Request a Quote
D13-9001 is a small-molecule efflux pump inhibitor used as a research tool to probe AcrB- and MexB-mediated antibiotic efflux. It is intended for laboratory research use only and is supplied as a solid with high reported purity and documented physicochemical properties.