Learn More
Medchemexpress LLC HY-50938 50mg Medchemexpress, D149 Dye CAS:786643-20-7 Purity:>98%
SDP
Catalog No. 502082260
Encompass_Preferred
Medchemexpress, HY-50938 50mg D149 Dye CAS:786643-20-7 D149 Dye is an indoline-based dye, which is a high-extinction-coefficient metal-free organic sensitizer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2260 Supplier Medchemexpress LLC Supplier No. HY5093850MG
Medchemexpress, HY-50938 50mg D149 Dye CAS:786643-20-7 D149 Dye is an indoline-based dye, which is a high-extinction-coefficient metal-free organic sensitizer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.