Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D159687 | 1155877-97-6 | C21H19ClN2O2 | 50 MG
SDP

Catalog No. 5000376802
Encompass_Preferred

D159687 is a selective PDE4D inhibitor. It is for research use only and not sold to patients. This compound has a molecular weight of 366.84 and appears as a white to off-white solid with a purity of 98.42%.

  • Selective PDE4D inhibitor
  • Molecular weight of 366.84
  • Solid appearance, white to off-white color
  • High purity of 98.42%
  • Soluble in DMSO at 150 mg/mL (408.90 mM)
  • Induces a transient increase in CREB phosphorylation in HT-22 cell line
  • Demonstrates potential for recruitment or enhancement of synaptic function in female Cynomolgus macaques

Catalog No. 50-003-76802 Supplier Medchemexpress LLC Supplier No. HY1544450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D159687 is a selective PDE4D inhibitor. It is for research use only and not sold to patients. This compound has a molecular weight of 366.84 and appears as a white to off-white solid with a purity of 98.42%.

  • Selective PDE4D inhibitor
  • Molecular weight of 366.84
  • Solid appearance, white to off-white color
  • High purity of 98.42%
  • Soluble in DMSO at 150 mg/mL (408.90 mM)
  • Induces a transient increase in CREB phosphorylation in HT-22 cell line
  • Demonstrates potential for recruitment or enhancement of synaptic function in female Cynomolgus macaques

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.