Learn More
Aviva Systems Biology Corp D15Wsu75e Antibody - middle region (ARP46230_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46230_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against D15Wsu75e. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human D15Wsu75e. Alternate names: DJ347H13.4, MGC138384, FAM152B, D15Wsu75eApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59280