Learn More
Medchemexpress LLC D18 | 2230218-36-5 | 98.8% | 364.5 g/mol | C21H28N6 | 5 MG
SDP
Catalog No. 5000222596
Encompass_Preferred
D18 is a small-molecule TLR7/8 dual agonist and ADC cytotoxin used in immunology and oncology research. It activates TLR7 and TLR8, increases PD-L1 expression via epigenetic regulation, and is used as a building block for antibody-drug conjugate synthesis; supplied for laboratory research use only.
  • Dual TLR7/8 agonist activity.
  • Enhances PD-L1 expression through epigenetic regulation.
  • Acts as an ADC cytotoxin suitable for antibody-drug conjugate synthesis.
  • High reported purity (~98.8%).
  • Supplied in small milligram quantities; soluble in DMSO.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-22596 Supplier Medchemexpress LLC Supplier No. HY1445015MG
Add to Cart
Add to Cart
D18 is a small-molecule TLR7/8 dual agonist and ADC cytotoxin used in immunology and oncology research. It activates TLR7 and TLR8, increases PD-L1 expression via epigenetic regulation, and is used as a building block for antibody-drug conjugate synthesis; supplied for laboratory research use only.
  • Dual TLR7/8 agonist activity.
  • Enhances PD-L1 expression through epigenetic regulation.
  • Acts as an ADC cytotoxin suitable for antibody-drug conjugate synthesis.
  • High reported purity (~98.8%).
  • Supplied in small milligram quantities; soluble in DMSO.
  • Intended for research use only; not for human or clinical use.