Learn More
Analytical Sales & Services D2 Lamp for Beckman Model 163
Catalog No. 5000807258
Encompass
D2 Lamp for Beckman Model 163; Camag Model TLC II Scanner; Gilford Models 200 220 222 230 240 250 252 260 2000 2400 2600; Isco Model U4/1840; Waters Models 480 481
Catalog No. 50-008-07258 Supplier Analytical Sales & Services Supplier No. 12205
D2 Lamp for Beckman Model 163; Camag Model TLC II Scanner; Gilford Models 200 220 222 230 240 250 252 260 2000 2400 2600; Isco Model U4/1840; Waters Models 480 481