Learn More
Analytical Sales & Services D2 Lamp for Beckman Models DB
Catalog No. 5000804740
Encompass
D2 Lamp for Beckman Models DB DBG DBGT DK DU; Shimadzu Models SP-4 SPD-2A 2AV 6A 6AV; Varian/Cary/Rainin Models LC-5000 5500 UV50 100 200 PRO STAR 310 STAR 9050 VISTA 5500
Qty/Pkg: EACH
Catalog No. 50-008-04740 Supplier Analytical Sales & Services Supplier No. 12305
D2 Lamp for Beckman Models DB DBG DBGT DK DU; Shimadzu Models SP-4 SPD-2A 2AV 6A 6AV; Varian/Cary/Rainin Models LC-5000 5500 UV50 100 200 PRO STAR 310 STAR 9050 VISTA 5500
Qty/Pkg: EACH