Learn More
Analytical Sales & Services D2 Lamp & Holder for Applied
Catalog No. 5000806559
Encompass
D2 Lamp & Holder for Applied Biosystems/Kratos/Perkin Elmer Models 120A 130A 270A-HT 1000S FS970 FS980 SF757 SF773 SF783 SF783A SF785A SF769 SF770; Waters Model 450
Qty/Pkg: EACH
Catalog No. 50-008-06559 Supplier Analytical Sales & Services Supplier No. 1281
D2 Lamp & Holder for Applied Biosystems/Kratos/Perkin Elmer Models 120A 130A 270A-HT 1000S FS970 FS980 SF757 SF773 SF783 SF783A SF785A SF769 SF770; Waters Model 450
Qty/Pkg: EACH