Learn More
Analytical Sales & Services D2 Lamp & Holder for Models DU
Catalog No. 5000807054
Encompass
D2 Lamp & Holder for Models DU-60 62 63 64 65 68.
Qty/Pkg: EACH
Catalog No. 50-008-07054 Supplier Analytical Sales & Services Supplier No. 12871
D2 Lamp & Holder for Models DU-60 62 63 64 65 68.
Qty/Pkg: EACH