Learn More
Analytical Sales & Services D2 Lamp & Holder for Models UV
Catalog No. 5000807276
Encompass
D2 Lamp & Holder for Models UVD160 170S 170U 320 340S 340U.
Qty/Pkg: EACH
Catalog No. 50-008-07276 Supplier Analytical Sales & Services Supplier No. 12891
D2 Lamp & Holder for Models UVD160 170S 170U 320 340S 340U.
Qty/Pkg: EACH