Learn More
Analytical Sales & Services D2 Lamp & Holder for Perkin-El
Catalog No. 5000806026
Encompass
D2 Lamp & Holder for Perkin-Elmer/ABI Models LC-480 Lambda 2 5 9 10 11 12 14 through 20 25 35 40 45; 650 800 850 900 950 BIO;\n\nQty/Pkg: EACH
Catalog No. 50-008-06026 Supplier Analytical Sales & Services Supplier No. 12892
D2 Lamp & Holder for Perkin-Elmer/ABI Models LC-480 Lambda 2 5 9 10 11 12 14 through 20 25 35 40 45; 650 800 850 900 950 BIO;\n\nQty/Pkg: EACH