Learn More
Analytical Sales & Services D2 Lamp & Holder for Perkin-El
Catalog No. 5000804794
Encompass
D2 Lamp & Holder for Perkin-Elmer/ABI Models 220 320 330 340 556 557 EZ150 201 210
Qty/Pkg: EACH
Catalog No. 50-008-04794 Supplier Analytical Sales & Services Supplier No. 12851
D2 Lamp & Holder for Perkin-Elmer/ABI Models 220 320 330 340 556 557 EZ150 201 210
Qty/Pkg: EACH