Learn More
Analytical Sales & Services D2 Lamp & Holder for Shimadzu
Catalog No. 5000806580
Encompass
D2 Lamp & Holder for Shimadzu Models LC2010AHT LC2010CHT
Qty/Pkg: EACH
Catalog No. 50-008-06580 Supplier Analytical Sales & Services Supplier No. 12884
D2 Lamp & Holder for Shimadzu Models LC2010AHT LC2010CHT
Qty/Pkg: EACH