Learn More
Azzota Corp Deuterium Lamp For Thermo Dionex Dad-3000(Rs), Mwd-3000(Rs) And Vwd-3X00(Rs) Series Detector, Vwd-3100, Vwd-3400, Dad-3400, Dad-3400Rs, Mwd-3000 And Other Hplc Uv Detector Instruments.
Catalog No. 502060348
Encompass_Preferred
Deuterium Lamp for Thermo Dionex DAD-3000(RS), MWD-3000(RS) and VWD-3x00(RS) Series Detector. For VWD-3100, VWD-3400, DAD-3400, DAD-3400RS, MWD-3000 and other HPLC UV detector lab instruments. Also compatible with Dionex ion chromatography UV VWD module. Compatible Thermo part number 6074.1110
Catalog No. 50-206-0348 Supplier Azzota Corp Supplier No. D2T6074
Deuterium Lamp for Thermo Dionex DAD-3000(RS), MWD-3000(RS) and VWD-3x00(RS) Series Detector. For VWD-3100, VWD-3400, DAD-3400, DAD-3400RS, MWD-3000 and other HPLC UV detector lab instruments. Also compatible with Dionex ion chromatography UV VWD module. Compatible Thermo part number 6074.1110