Learn More
Azzota Corp D2 Lamp For Waters Acquity Pda/Tuv 2489/2998 Perf Maint Kit, 2489 Dual Wavelength Uv Detector, 2998 Pda Detector, And Acquity Elambda Pda Detector, Equivalent To Waters Pn 201000281 Or 201000186
Catalog No. 501612823
Encompass_Preferred
D2 lamp for Waters ACQUITY PDA/TUV 2489/2998 PERF MAINT KIT. For 2489 Dual Wavelength UV Detector, the 2998 PDA Detector, and the ACQUITY eLambda PDA Detector. Equivalent to Waters part number 201000281 or 201000186
Catalog No. 50-161-2823 Supplier Azzota Corp Supplier No. D2WT0281
D2 lamp for Waters ACQUITY PDA/TUV 2489/2998 PERF MAINT KIT. For 2489 Dual Wavelength UV Detector, the 2998 PDA Detector, and the ACQUITY eLambda PDA Detector. Equivalent to Waters part number 201000281 or 201000186