Learn More
Alkali Scientific D2400-R BeadBlaster 24R Refrig
SDP
Catalog No. 503029521
Encompass_Preferred
Get out of the cold room with the new refrigerated Alkali ScientificD2400-R BeadBlaster24R This powerful bead homogenizer completely lyses grinds and homogenizes a broad variety of biological samples The refrigeration system protects heat sensitive samples during homogenization Even the most difficult and resistant samples are fully homogenized often in 35 seconds or less The stainless steel tube carrier subjects up to 24 samples to an optimized high speed 3D motion producing high energy impacts between the samples and micro-beads to release cellular contents After homogenization samples can be centrifuged and the supernatant collected for further processing Operating parameters for the Alkali ScientificD2400-R BeadBlaster 24R Refrigerated Bead Homogenizer are set and displayed digitally Up to 50 programs can be stored in memory and recalled for future use The mechanical lid lock prevents opening when the BeadBlaster 24 is in operation while the brushless motor reduces motor noi
Catalog No. 50-302-9521 Supplier Alkali Scientific Supplier No. D8400R
Add to Cart
Add to Cart
Request a Quote
Get out of the cold room with the new refrigerated Alkali ScientificD2400-R BeadBlaster24R This powerful bead homogenizer completely lyses grinds and homogenizes a broad variety of biological samples The refrigeration system protects heat sensitive samples during homogenization Even the most difficult and resistant samples are fully homogenized often in 35 seconds or less The stainless steel tube carrier subjects up to 24 samples to an optimized high speed 3D motion producing high energy impacts between the samples and micro-beads to release cellular contents After homogenization samples can be centrifuged and the supernatant collected for further processing Operating parameters for the Alkali ScientificD2400-R BeadBlaster 24R Refrigerated Bead Homogenizer are set and displayed digitally Up to 50 programs can be stored in memory and recalled for future use The mechanical lid lock prevents opening when the BeadBlaster 24 is in operation while the brushless motor reduces motor noi