Learn More
Medchemexpress LLC D2A21 TFA | >95.6% | 2775.42 | 1 MG
SDP
Catalog No. 5000423429
Encompass_Preferred
D2A21 TFA is an antimicrobial peptide that exhibits significant activity against Pseudomonas and has a bactericidal effect on burn scabs. This peptide also demonstrates antitumor activity, making it a candidate for research in prostate cancer.
  • Antimicrobial peptide
  • Active against Pseudomonas
  • Bactericidal effect on burn scabs
  • Antitumor activity
  • Used in prostate cancer research
Catalog No. 50-004-23429 Supplier Medchemexpress LLC Supplier No. HYP11211A1MG
D2A21 TFA is an antimicrobial peptide that exhibits significant activity against Pseudomonas and has a bactericidal effect on burn scabs. This peptide also demonstrates antitumor activity, making it a candidate for research in prostate cancer.
  • Antimicrobial peptide
  • Active against Pseudomonas
  • Bactericidal effect on burn scabs
  • Antitumor activity
  • Used in prostate cancer research