Learn More
Cayman Chemical D2PM (hydrochloride), Purity: ≥98%, CAS: 172152-19-1, 50 mg
SDP
Catalog No. 502955846
Encompass_Preferred
A psychoactive designer drug; also used in organic synthesis to prepare the Corey-Bakshi-Shibata catalyst; intended for forensic purposes
Catalog No. 50-295-5846 Supplier Cayman Chemical Supplier No. 1116050 MG
A psychoactive designer drug; also used in organic synthesis to prepare the Corey-Bakshi-Shibata catalyst; intended for forensic purposes