Learn More
Medchemexpress LLC D3/5-HT receptor modulator-1 | 898532-85-9 | 391.51 | 1 ML
SDP
Catalog No. 5000346130
Encompass_Preferred
D3/5-HT receptor modulator-1 (compound 5i) is a selective dopamine D3 receptor and 5-HT2A receptor antagonist and a partial 5-HT1A receptor agonist. It has a low affinity for dopamine D2 receptors, 5-HT2C receptors, and hERG channels, and presents an atypical antipsychotic profile.
  • Selective dopamine D3 receptor and 5-HT2A receptor antagonist.
  • Partial 5-HT1A receptor agonist.
  • Shows Ki values of 4.5 nM (D3), 11.9 nM (5-HT2A), and 15.3 nM (5-HT2A).
  • Low affinity for dopamine D2 receptors, 5-HT2C receptors, and hERG channels.
  • Atypical antipsychotic profile.
Catalog No. 50-003-46130 Supplier Medchemexpress LLC Supplier No. HY1086110MG
D3/5-HT receptor modulator-1 (compound 5i) is a selective dopamine D3 receptor and 5-HT2A receptor antagonist and a partial 5-HT1A receptor agonist. It has a low affinity for dopamine D2 receptors, 5-HT2C receptors, and hERG channels, and presents an atypical antipsychotic profile.
  • Selective dopamine D3 receptor and 5-HT2A receptor antagonist.
  • Partial 5-HT1A receptor agonist.
  • Shows Ki values of 4.5 nM (D3), 11.9 nM (5-HT2A), and 15.3 nM (5-HT2A).
  • Low affinity for dopamine D2 receptors, 5-HT2C receptors, and hERG channels.
  • Atypical antipsychotic profile.