Learn More
Medchemexpress LLC D3/5-HT receptor modulator-1 | 898532-85-9 | 391.51 | 25 MG
SDP
Catalog No. 5000347090
Encompass_Preferred
D3/5-HT receptor modulator-1 (compound 5i) is a selective dopamine D3 receptor and 5-HT2A receptor antagonist, and a partial 5-HT1A receptor agonist. This modulator has an atypical antipsychotic profile.
  • Shows Kᵢ values of 4.5 nM, 11.9 nM, and 15.3 nM for dopamine D3, 5-HT2A, and 5-HT1A receptors, respectively.
  • Has low affinity for dopamine D2 receptors, 5-HT2C receptors, and hERG channels.
Catalog No. 50-003-47090 Supplier Medchemexpress LLC Supplier No. 25MGHY10861
Add to Cart
Add to Cart
D3/5-HT receptor modulator-1 (compound 5i) is a selective dopamine D3 receptor and 5-HT2A receptor antagonist, and a partial 5-HT1A receptor agonist. This modulator has an atypical antipsychotic profile.
  • Shows Kᵢ values of 4.5 nM, 11.9 nM, and 15.3 nM for dopamine D3, 5-HT2A, and 5-HT1A receptors, respectively.
  • Has low affinity for dopamine D2 receptors, 5-HT2C receptors, and hERG channels.