Learn More
Medchemexpress LLC D3/5-HT receptor modulator-1 | 898532-85-9 | 99.8% | 391.51 g·mol⁻¹ | C24H29N3O2 | 5 MG
SDP
Catalog No. 5000263800
Encompass_Preferred
D3/5-HT receptor modulator-1 (compound 5i) is a small-molecule research reagent with a mixed dopaminergic and serotonergic profile. It functions as a selective dopamine D3 receptor antagonist, a 5-HT2A receptor antagonist, and a partial 5-HT1A receptor agonist, and is intended for in vitro and preclinical studies investigating receptor pharmacology and antipsychotic-like mechanisms.
  • Selective dopamine D3 receptor antagonist, 5-HT2A receptor antagonist, and partial 5-HT1A receptor agonist.
  • High purity (≈99.8%).
  • Molecular formula C24H29N3O2 and molecular weight 391.51 g·mol⁻¹.
  • Suitable for in vitro and preclinical receptor pharmacology studies.
  • Available in small milligram packages for assay screening.
Catalog No. 50-002-63800 Supplier Medchemexpress LLC Supplier No. HY108615MG
D3/5-HT receptor modulator-1 (compound 5i) is a small-molecule research reagent with a mixed dopaminergic and serotonergic profile. It functions as a selective dopamine D3 receptor antagonist, a 5-HT2A receptor antagonist, and a partial 5-HT1A receptor agonist, and is intended for in vitro and preclinical studies investigating receptor pharmacology and antipsychotic-like mechanisms.
  • Selective dopamine D3 receptor antagonist, 5-HT2A receptor antagonist, and partial 5-HT1A receptor agonist.
  • High purity (≈99.8%).
  • Molecular formula C24H29N3O2 and molecular weight 391.51 g·mol⁻¹.
  • Suitable for in vitro and preclinical receptor pharmacology studies.
  • Available in small milligram packages for assay screening.