Learn More
Medchemexpress LLC HY-124867 5mg Medchemexpress, D3-βArr CAS:662164-09-2 Purity:>98%
SDP
Catalog No. 501969684
Encompass_Preferred
Medchemexpress, HY-124867 5mg D3-βArr CAS:662164-09-2 D3-βArr is a positive allosteric modulator for thyrotropin receptor (TSHR), which initiates translocation of β-Arr 1 by direct TSHR activation and potentiates TSH-mediated preosteoblast differentiation in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9684 Supplier Medchemexpress LLC Supplier No. HY1248675MG
Medchemexpress, HY-124867 5mg D3-βArr CAS:662164-09-2 D3-βArr is a positive allosteric modulator for thyrotropin receptor (TSHR), which initiates translocation of β-Arr 1 by direct TSHR activation and potentiates TSH-mediated preosteoblast differentiation in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.