Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D3-βarr | 662164-09-2 | 99.5% | 333.43 | C20H23N5 | 10MG
SDP

Catalog No. 5000239320
Encompass_Preferred

D3-βArr is a research-grade TSH receptor modulator used to probe receptor signaling and positive allosteric modulation in vitro. It is supplied as a high-purity solid with characterized molecular data for use in biochemical and cell-based assays.

  • High purity (~99.5%) suitable for research applications.
  • Molecular weight 333.43 g/mol.
  • Chemical formula C20H23N5.
  • Supplied as a solid in small-milligram quantities for screening.
  • Intended for in vitro biochemical and cell-based assays.
  • Not for human or clinical use; for research use only.

Catalog No. 50-002-39320 Supplier Medchemexpress LLC Supplier No. HY12486710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D3-βArr is a research-grade TSH receptor modulator used to probe receptor signaling and positive allosteric modulation in vitro. It is supplied as a high-purity solid with characterized molecular data for use in biochemical and cell-based assays.

  • High purity (~99.5%) suitable for research applications.
  • Molecular weight 333.43 g/mol.
  • Chemical formula C20H23N5.
  • Supplied as a solid in small-milligram quantities for screening.
  • Intended for in vitro biochemical and cell-based assays.
  • Not for human or clinical use; for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.