Learn More
Medchemexpress LLC D3-βArr | 662164-09-2 | 99.5% | 333.43 | C20H23N5 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12486725MG
Encompass_Preferred
D3-βArr is a small-molecule positive allosteric modulator of the thyrotropin (TSH) receptor supplied for research use. It is provided as a solid powder and as a 10 mM solution in DMSO for in vitro studies, and has defined molecular properties and high purity suitable for biochemical and cell-based assays.
  • Positive allosteric modulator of the thyrotropin (TSH) receptor
  • High purity (99.5%) supporting reproducible results
  • Molecular formula C20H23N5; molecular weight 333.43
  • Available as solid (5 mg, 10 mg, 25 mg, 50 mg) and 10 mM solution in DMSO
  • Suitable for biochemical and cell-based assay applications
  • For research use only; not for human or veterinary use
Catalog No. 50-002-38828