Learn More
SPECTRUM QUALITY STANDARDS D3606 Check Standards – +4% 2-Butanol – 10 day set
Supplier:  SPECTRUM QUALITY STANDARDS 3606D3B10
Encompass_Preferred
D3606 Check Standards - +4% 2-Butanol - 10 day set. Accuracy of the Calibration can be monitored daily by the use of Check Standards.
Catalog No. NC1016090